The Active view displays the active systems in a laboratory through system cards. Each card represents a system that contains individual instruments. The individual instruments of a system are displayed on that system's card as tiles. The following table describes the parameters that are available on a system card on the Active view.
Function | Subfunction | Description |
|---|---|---|
System Name | N/A | Navigates to the system profile page. (For Chromeleon CDS) Specifies the name of the instrument provided during configuration. NOTE The system name is not editable in the Instrument application. When the system name is duplicated or truncated, pointing to it displays the full system name along with the instrument PC name. (For Xcalibur data system) Specifies the instrument PC name and the timestamp. When the system name is truncated, pointing to it displays the full system name. |
Instrument tile | N/A | Navigates to the instrument profile page. |
| N/A | Specifies a new instrument notification. |
| N/A | Specifies that a service ticket is opened on this instrument. |
| N/A | Specifies that an instrument is marked as a favorite. |
| N/A | Specifies a change in the system configuration. |
Connection Lost | System with instrument connectivity issues | Specifies that the connection to the Ardia Platform Link on the instrument PC is lost. |
Injection | System with real-time acquisition | Displays the active injection details and the injection number. |
Sequence | System with real-time acquisition | Displays the sequence name of the active sequence and the time remaining for the current sequence. |
Queue | System with real-time acquisition | (For Chromeleon CDS) Displays the total number of sequences in a queue. (For Xcalibur data system) Displays the total remaining number of sequences in a queue. |
No Upcoming Reservations | System with no upcoming reservations | Specifies that the system has no upcoming reservations and it is available to use. |
Go to Scheduler | System with no upcoming or active reservations | Navigates to the Scheduler page. |
Current Reservation | System with active reservations | Displays the current reservation details, such as the reservation name, reservation assignee, and the end date and end time of the reservation. |
Current Reservation for Maintenance | System with active reservations | Displays the current maintenance reservation details, such as the reservation name, reservation assignee, and the end date and end time of the reservation. |
Upcoming Reservation | System with upcoming reservations | Displays the upcoming reservation details, such as the reservation name, reservation assignee, and the start date and start time of the reservation. |
Upcoming Reservation for Maintenance | System with upcoming reservations | Displays the upcoming maintenance reservation details, such as the reservation name, reservation assignee, and the start date and start time of the reservation. |