The Systems page displays the entire fleet of systems in the laboratory. This page serves as a dashboard that consists of cards. Each card represents a system (LC, GC, IC, LC/MS, GC/MS, or IC/MS). By default, the application opens to the Systems view. The following table describes the functions on the Systems page.
Function | Description |
|---|---|
Active | Displays the active connected systems that are synced from the Xcalibur software or the Chromeleon CDS. Allows users to navigate to the profiles of systems and individual instruments. |
Disconnected | Displays instruments that were previously connected and active but are currently unconfigured or disconnected from the system. |
Waiting for Setup | Displays systems that are configured in the Xcalibur software or the Chromeleon CDS. To access further details of a system, the setup process needs to be completed. |
Welcome banner | Includes a link to the Ardia Instruments Application Quick Start Guide. |