Software section | Abstract |
|---|---|
Instruments | The Ardia Instruments application only supports instrument notifications from the Vanquish Horizon modules. |
Systems | Chromeleon CDS users must save the Instrument Configuration within the software to ensure the configuration details are synchronized with the Ardia Instruments application. |
Systems | Chromeleon software users can only view the chromatogram display on the system card for the current default channel. |
Systems | If a user connects the Instrument PC to the Ardia Platform through the Ardia Platform Link application, it might take up to one to two minutes to view the Waiting for Setup page. |