The following table lists known defects in the software with a brief abstract about the defect and information related to each defect’s severity and risk. The Item ID corresponds to the internal number assigned to each issue. Product management assesses the risk, which might differ significantly from the reported severity.
Software section
Item ID
Abstract
Severity
Risk
Systems
365457
Sometimes, the System card fails to display the complete chromatogram when it has significantly delayed eluting peaks.
Medium
Medium
Systems
366238
The number of sequences displayed on the System card represents the total count of sequences in the queue (Chromeleon CDS acquisition).
Low
Low
Systems
381930
The System card for the disconnected instruments does not display the current or upcoming scheduler reservations.
Low
Low
Systems
383258
The sequence information is not displayed on the System card when the queue is stopped after the injection or stopped after the sequence.
Medium
Low
Systems
331031
The disconnected tab does not support the filtering of instruments.
Low
Low
Instrument connectivity
343958
The creation of instruments with identical names within a Chromeleon CDS installation is not supported.
High
Medium
Scheduler
375802
When the assignee and the initiator are the same user, the notifications are duplicated and sent twice.
Low
Low
Instrument connectivity
402801
After restarting the Ardia Platform Link services, the active system card might not display the chromatogram and the remaining sequence time. In this case, restart the services again.
Low
Low