The user cannot initiate a support request for an instrument if the instrument's profile does not contain service provider details. In such cases, selecting Request Support opens the Request Support Troubleshooting dialog box with one of the messages listed in the following table.
  • Service provider details might be missing from the system information.
  • The SN is missing from the system information.
  • The application is unable to find your devices in our inventory table.
  • Connection interrupted.