The Waiting for Setup view displays all the systems that are connected to the Ardia Platform through the Xcalibur software or the Chromeleon CDS. Users can edit or enter the functions described in the following table.
Function | Subfunction | Description |
---|---|---|
Complete Setup | N/A | Adds or modifies the instrument attribute details and initiates real-time monitoring. |
System Setup | System Name | Displays the name of the system. NOTE This field is editable only for a system configured in the Xcalibur data system. |
Instrument Name | Displays the name of the instrument. The default name comes from the detected instrument configuration. Users can edit this name, which can be up to 40 characters long and include alphanumeric characters and special characters (for example: _, /, -). | |
Serial Number (SN) | Displays the SN of the instrument, which can be read-only or editable. The SN cannot be edited if the SN field is prefilled with the SN linked to the registered instrument. | |
Location | Specifies the location of the instrument. | |
Room | Specifies the room number where the instrument is located. | |
Owners | Specifies the owners of the instrument and sends the instrument notifications via email. Multiple owners can be assigned to an instrument. | |
Custom Tag | Specifies the custom tags for the system or instrument. | |
NOTE If the lab is connected to Thermo Fisher and the device is under contract, the application displays the Service Contact dialog box. | ||
Service Contact dialog box (device under contract) | N/A | Specifies the name of the contact person selected in the organization for the Unity Lab Services to use as the primary contact. NOTE Only applicable if Unity Lab Services is used for instrument service. |
Is this instrument on an asset management plan? | Yes | Specifies whether the instrument is on asset management services. NOTE The Request Support button is not available for the instrument. |
No | Allows users to add the required contact details (username, email, and phone number). NOTE The Request Support button is available for the instrument. | |
NOTE If the lab is connected to Thermo Fisher and the device is not under contract, the application displays the Service Contact dialog box. | ||
Service Contact dialog box (device not under contract) | N/A | Specifies the name of the contact person selected in the organization for the Unity Lab Services to use as the primary contact. NOTE Only applicable if Unity Lab Services is used for instrument service. |
Do you contact Unity Lab Services for service? | Yes | Allows users to add the required contact details (username, email, and phone number). NOTE The Request Support button is available for the instrument. |
No | Specifies the instrument is on system management services. NOTE The Request Support button is not available for the instrument. |