You can add the labels for the mass spectrum peaks from the Display Options dialog box.
Procedure
- Open an injection or a raw data file (see Open injections or raw data files.
- The Chromatogram view and Spectrum view open.
- Select the Spectrum view.
- Select the Ellipsis menu
to open the Display Options dialog box.
- In the Labels area, select the labels that you want to use to annotate the spectrum peaks (see Options for peak labels).
TIP
To avoid labeling low-intensity mass peaks, specify a higher relative intensity value in the Label Threshold box. When you set the default value to 0 in the Label Threshold box, the application labels all the peaks.
Mass spectrum with multiple labels and a labeling threshold of 50% relative intensity