You can add an XIC trace to the Chromatogram view using the Spectrum view.
Procedure
- Select the Spectrum view.
- In the Spectrum view, point to the m/z label that you want to use for the XIC trace.
- Double-click the m/z value to create an XIC trace from the selected mass.
- A new XIC trace appears at the bottom of the Chromatogram view, and the selected mass appear in the Ranges column of the Ranges dialog box.
| No. | Description | No. | Description |
|---|---|---|---|
1 | XIC trace | 2 | m/z of XIC trace |
NOTE
To undo all changes made to this page and to revert to the previously applied changes, select .