An active workspace is saved when you exit or sign out from the Data Viewer application or the Ardia platform.
NOTE
A workspace is saved only when a function is performed in the active tab.
Procedure
- Open the raw data file of interest.
- The Chromatogram view and Spectrum view open.
- Perform any function in the active workspace. For example, modifications to the Spectrum view and the Chromatogram view (such as performing spectrum averaging, modifying peak detection settings, changing chromatogram ranges, labeling peaks, processing data, and so on).
- Do one of the following:
- Sign out from the Ardia platform.
NOTE
When you navigate away from the Data Viewer or sign out of Ardia platform, certain changes made to your active workspace are saved. This includes modifications to the Spectrum view and Chromatogram view.
- Exit the application by selecting Thermo Scientific Ardia Platform
NOTE
The active workspace does not save if you select the back button, refresh the page, change the URL, or visit a bookmark page.
- A confirmation prompt to save the workspace opens.
- Select Yes to save the active workspace, or select No to exit the application without saving the workspace.