In the Data Viewer workspace, you can open a single injection or raw data file, or you can open multiple injections or raw data files from the Ardia repository.
Procedure
- Do one of the following:
- In the Data Viewer application menu bar, select File > Open.
- Select
.
- The Ardia repository folder opens.
- Select the checkboxes of one or more injections or raw data files that you want to open, then select Open.
- A workspace appears for each injection or raw data file (Display a workspace).