You can add Precursor Flag markers and Nearby Precursors markers to spectrum peaks. Double-clicking these markers opens the data-dependent scans in another view.
Procedure
- Double-click on the precursor marker to display its product ion spectrum in a separate Spectrum view.
| No. | Description | No. | Description |
|---|---|---|---|
1 | Precursor marker on the m/z 195.0877 peak in the full MS1 scan | 3 | Precursor marker on the m/z 110.0713 peak |
2 | Data-dependent MS2 scan for m/z 195.0877 | 4 | Precursor marker on the m/z 138.0662 peak |
The following figures show the product ion scans for the two marked precursor ions in the data-dependent MS2 scan.