Peak labeling customizes the display of the selected Spectrum view. Use the Labels area in the Display Option dialog box to add or remove labels that appear above the spectrum peaks. By default, the application labels each mass spectrum peak with its corresponding m/z value. The following table shows different options for peak labels.
Option | Description |
|---|---|
Mass | Adds an m/z label to each peak in the spectrum. |
Flags | Displays one of the following letters or symbols for the flagged peaks:
|
Resolution | Adds a peak resolution label to each peak in the spectrum. The abbreviation R next to a value indicates the peak resolution. |
Charge | Adds a charge state label to each peak in the spectrum. The abbreviation z next to a value indicates the charge state. |
Baseline | Adds a baseline label to each peak in the spectrum. The abbreviation B next to a value indicates the baseline. |
Noise | Adds a noise magnitude label to each peak in the spectrum. The abbreviation N next to a value indicates the noise. |
Width (m/r) | Adds a width label to each peak in the spectrum. The width equals the mass, in mmu units, divided by the peak resolution. The abbreviation W next to a value indicates the width label. |
Monoisotopic Mass | Displays the monoisotopic mass and peak label (MM) in the Spectrum view.
|
Species ID | Displays a species ID label (S) above mass spectral peaks that belong to the same chemical species (with the same species ID). The application does not label peaks with a species ID label if they do not have an assigned species ID. |
Reference and Exception Peaks | Shows or hides the reference peaks and exception peaks in the Spectrum view. |
Precursor Flag | A marker ( |
Nearby Precursor | A marker ( In the Nearby Precursor area, you can specify the search for the data-dependent scans using the following options:
|
Label Threshold | Sets the percentage of the base peak so that the application labels only the peaks above that percentage. For example, if the base peak is 100 percent and the label threshold setting is 50 percent, the application labels only the peaks in the Spectrum view or MultiSpectrum view that are at or above 50 percent. Range: 0–100; default: 0% |