When a spectrum is linked to a chromatogram trace, you can use the Average Spectrum ellipsis menu to create an average spectrum.
Procedure
- Open an injection or a raw data file.
- Select the Chromatogram view.
- Select
, and Average Spectrum option from the dropdown list.
- The cursor changes to the averaging cursor,
.
- Create an averaged spectrum by using any of the following procedures:
- Select the RT range using the cursor
- Edit the averaging range from the dialog box