You can create an averaged spectrum by using the Processing toolbar.
Procedure
- Open an injection or a raw data file.
- Select the Chromatogram view.
- Select the Processing tab to display the Processing Options for Chromatogram view.
Processing options for a chromatogram - Select Average Spectrum
to make the average spectrum options available.
- The cursor changes to the averaging cursor,
.
- Follow any one of the procedures:
- Manually entering RT range and scan range (see Enter the RT range or scan range)
- Create RT range using the cursor (see Select the RT range using the cursor)
- Entering RT range an scan range in the dialog box (see Edit the averaging range from the dialog box)