You can export the Chromatogram view by downloading it as an SVG file, or you can copy the view to the clipboard and then paste it into a document.
Procedure
- Select
, and then select the Export as Image option from the dropdown menu.
- The Export as Image dialog box opens.
Export as Image dialog box - In the Export as Image dialog box, select one of these options:
- To copy the active Chromatogram view to the clipboard, select the Copy to Clipboard option.
- The Chromatogram view is copied and you can paste the image in the appropriate document.
- To download the active Chromatogram view as an SVG image, do the following:
- Select the Download as SVG option.
- Enter the width and height of the image in the corresponding boxes in the Dimensions area.
- The valid range is 2 to 50. The units of the value are either millimeters or inches, which depends on the units option that you select.
- Select Export.
- Depending on the export type, do one of the following:
- For the Copy to Clipboard option, paste the copied Chromatogram view into a document.
- For the Download as SVG option, select the folder location where you want to save the image, name the file, and then select Save. You can then insert the image from the SVG file into a document.