The default values on the Peak Detection page are suitable for most analysis requirements. Change these settings only if standard chromatogram detection and integration options do not provide the desired result.
Procedure
  1. Open a raw data file.
  2. Expand the left sidebar, and then select the Peak Detection tab.
  3. Change the settings as appropriate. For more information, see Select the manual noise region