You can select scan filters to specify the process that the application applies to a subset of the scans in a raw data file. Use only the fields that apply to your mass spectrometer. In the Chromatogram view (see Modify Chromatogram view), you can define additional scan filters by adhering to the following scan filter formats.
NOTE
Not all features are available for every mass spectrometer.
The following table lists the scan filter formats.
Scan filter formats
Feature
Option
Interpretation
Polarity
+, –
Positive or negative.
Data type
p, c
Profile or centroid.
Dependent scans
d, !d
Include dependent scans or exclude dependent scans.
TurboScans
t, !t
Include TurboScan scans or exclude TurboScan scans.
Source CID
cid, !cid
Include Source CID scans or exclude Source CID scans.
(Source CID scans are scans for ions that are produced by collision-induced dissociation in the ion source.)
Scan type
FULL
Full scan
Z
ZoomScan
SIM
Selected ion monitoring
SRM
Selected reaction monitoring
CRM
Consecutive reaction monitoring
Q1MS
Q1 quadrupole analysis
Q3MS
Q3 quadrupole analysis
Scan mode
ms, ms2, ms3, ... ms10
MSn , where n = 1 to 10
Each order can be followed by the appropriate number of parent ions. The parent ions can also be omitted.
Example: “ms3 345.3, 253.2” indicates an MS3 scan with parent ions whose m/z values are 345.3 and 253.2.
pr
Parent ion (followed by the product ion mass)
cng
Constant neutral gain (followed by the mass that is being gained)
cnl
Constant neutral loss (followed by the mass being lost)
Mass Analyzer
ITMS
Ion trap
SQMS
Single-quadrupole
TQMS
Triple-quadrupole
FTMS
Fourier transform
Sector
Magnetic sector mass spectrometry
Photo Ionization
pi, !pi
Include photo ionization scans or exclude photo ionization scans.
Compensation Voltage
cv, !cv
Include compensation voltage scans or exclude compensation voltage scans.
Detector Valid
det, !det
Include detector valid scans or exclude detector valid scans.
Enhanced
E, !E
Include enhanced scans or exclude enhanced scans.
Wideband
w, !w
Include wideband scans or exclude wideband scans.
Supplemental Activation
sa, !sa
Include supplemental activation scans or exclude supplemental activation scans.
Multistate Activation
msa, !msa
Include multistate activation scans or exclude multistate activation scans.
Product masses or mass range of scan
[m1a–m1b, m2a–m2b, m3a–m3b, ...]
Scans with a specific mass range or mass ranges.
Example: "[50.00–1500.00]" represents a scan from an m/z of 50.00 to an m/z of 1500.00
If a scan is exactly 1 Da wide, it appears as a single value (the center mass). This is typical for SIM, SRM, and CRM. Filters for precursor ions in dependent scans are matched with a tolerance that is equal to an m/z of 1.0; this is so that so that minor differences in precursor mass measurements from scan to scan do not give different filters.
Segment/scan event number pairs
{segment, scan number}
Example: “{3, 4} + c ms” indicates segment 3, scan event 4 for a positive centroid MS scan
The braces, { }, are required.
Ionization mode
APCI
Atmospheric pressure chemical ionization
ESI
Electrospray
Example: “+ c ESI ms” indicates a positive centroid electrospray MS scan.
EI
Electron ionization
CI
Chemical ionization
NSI
Nanoelectrospray ionization
FAB
Fast atom bombardment
TSP
Thermospray ionization
FD
Field desorption
MALDI
Matrix-assisted laser desorption ionization
GD
Glow discharge
PSI
Paper spray ionization
Corona on/off
corona, !corona
Corona on or corona off.
Example: “+ APCI !corona ms” indicates a positive centroid APCI scan with the corona off.
Detector value
“det=##.##”
Detector value is ##.## with no spaces.
Example: “+ ESI det= –800.0” indicates a positive electrospray scan at –800.0 detector units (usually volts).
MS/MS and MSn CID energies
mass@energy
Mass is the precursor mass and energy is the CID relative energy (no units).
Example: “– c ms2 196.1@25.0” indicates a negative centroid MS/MS scan of m/z 196.1 at 25.0 units of CID energy.
Fragmentation/ disassociation technique
UVPD
Ultra violet photo dissociation
Quadrupole identification
Q1MS, Q3MS
Q1 quadrupole or Q3 quadrupole.
Example: “+ c ESI Q3MS” indicates a positive centroid electrospray MS scan using a Q3 quadrupole.
Accurate mass
AM, !AM
Include accurate mass scans, exclude accurate mass scans.
AMI, AME
Include accurate mass internal, or include accurate mass external.
Ultra
u, !u
Include ultra scans or exclude ultra scans.
Sector
BSCAN, !BSCAN
Include magnetic sector scans or exclude magnetic sector scans.
ESCAN, !ESCAN
Include electric sector scans or exclude electric sector scans.
LOCK
lock, !lock
Include lock scans or exclude lock scans.
Multiplex
msx, !msx
Include multiplexing scans or exclude multiplexing scans.
Electron capture dissociation
ecd, !ecd
Include electron capture dissociation or exclude electron capture dissociation.
Multi-photon dissociation
mpd, !mpd
Include photo dissociation or exclude photo dissociation.
Pulsed dissociation
pqd, !pqd
Include pulsed dissociation scans or exclude pulsed dissociation scans.
Electron transfer dissociation
etd, !etd
Include electron transfer dissociation scans or exclude electron transfer dissociation scans.
NPTR
nptr, !nptr
Include NPTR, or exclude NPTR.
Higher-energy CID
hcd, !hcd
Include higher energy CID scans or exclude higher energy CID scans.
Source SID
sid, !sid
Include source SID scans or exclude source SID scans.
(Source SID scans are surface-induced scans.)
Isolation width
iw ##, ##
Isolation width value at ##, ##.