The application records the scan method that you used as a scan event. The data system creates scan filters from the scan-event settings. You can select or create scan filters to apply processing to a subset of the scans in a raw data file. Scan headers provide important information about the scan. See these topics for more details: