Create an averaged spectrum by dragging the cursor across the x axis to select an RT range.
Procedure
- Enable the average spectrum from the Processing toolbar (see Average a spectrum from the Processing toolbar ) or the Ellipsis menu
(see Average a spectrum from the Ellipsis menu).
- Drag the pointer across the time range in the selected trace.
- A red line in the Chromatogram view marks the selected range and the Spectrum plot is updated with the averaged scan data. The RT range and the scan range are automatically updated in the range boxes irrespective of the selected option.
NOTE
To undo all changes made to this page and to revert to the previously applied changes, select . The Undo button is active only after applying changes to the averaged scan data.