To minimize the display of infrequently used information, some of the table columns in a view or dialog box are hidden. You can choose which columns display in a specific table by accessing the table’s Column dialog box.
Procedure
- To access the Columns commands do one of the following:
- In the Info Bar, select Ranges and then select Column.
- In Peak List view (Display Peak List view), select Column.
- Do any of the following:
- To show a column, select the associated checkbox.
- To hide a column, clear the associated checkbox.
NOTE
You can rearrange the rows of a column by pressing and holding
next to the row that you want to move, and then dragging the row up or down. The Column dialog box represent the columns in the Ranges table or Peak List view table.
- The following figure shows the Columns dialog box for the Peak List view.