Procedure
  1. Run a peak detection algorithm (see Display the Peak Detection page).
  2. In the Display Options for Chromatogram toolbar, select the options for the types of labels that you want displayed for the peaks in your chromatogram (see Label chromatogram peaks).
  3. (Optional) Clear the associated checkbox to remove an applied peak label from the Chromatogram view.