You can zoom in on and reset a Chromatogram view using the Zoom Range option in the Processing menu.
Procedure
- Select the Chromatogram view.
- Select the Processing tab.
- Select the Zoom Range option.
- Enter an RT range within the chromatogram RT limits.
- Press ENTER.
- The view rescales to display the chromatogram within the specified time range and relative abundance range.
- ( Optional) To return to the default scale, select Reset Zoom .
NOTE
To undo all changes made to this page and to revert to the previously applied changes, select .