You can change the injection or raw data file of the selected trace in the Ranges dialog box.
Procedure
- Select a trace in the Ranges dialog box.
- Select Browse
to the right of the Injection/Raw Data box in the Plot Properties and Processing area.
- The Data Explorer dialog box opens.
- Navigate to the Ardia repository folder for the required sequence (.seq), select an injection or a raw data file, and then select Open.
- To apply the changes to the active chromatogram, select Apply.
- The trace type depends on the experimental data in the selected injections or raw data files.
- (Optional) Close the Ranges dialog box as follows:
- Select Cancel or
.
- At the prompt that opens, do one of the following:
- Select Confirm to save your changes and close the dialog box.
- Select Cancel to close the dialog box without saving your changes.