Parallel reaction monitoring (PRM) acquires MS/MS scans based on a specified inclusion list (list of ions to be fragmented). This mode is most often used for small molecule applications involving analyses of complex sample matrices, which require ultimate selectivity.