SRM monitors a particular transition or set of transitions, such as the fragmentation of an ion or the loss of a neutral moiety. SRM monitors a limited number of precursor/product ion pairs. SRM provides the very rapid analysis of trace components in complex mixtures.