An MSn scan typically involves 3 to 10 stages of mass analysis (scan power of n = 3 to n = 10). Each stage of mass analysis where n > 2 includes an ion selection step. As you raise the scan power, you can obtain more structural information that can be useful in the structural elucidation of compounds of interest.