Mass spectra acquired with a high-resolution accurate-mass (HRAM) spectrometer include isotope clusters for analytes with elements that have more than one isotope. The mass difference between the isotopic peaks is proportional to the mass difference between the isotopes, and the relative intensity of the isotopic peaks is proportional to the natural abundance of the isotopes.