New features - New features - Ardia Data Viewer Application 1.1 Release Notes - Ardia Data Viewer - Ardia Data Viewer Application 1.1 Release Notes - New features - Ardia Data Viewer 1.1 Application Release Notes - Features - Ardia Data Viewer - topic
Ardia Data Viewer Application 1.1 Release Notes
- Product Tree
- Ardia Platform > Ardia Core Applications > Ardia Data Viewer
- Document Type
- Release Notes
- Software version
- 1.1
- A new opening screen to quickly open injections, restore a previous workspace, or access Help documents.
- Added right-click menus to the chromatogram and spectrum plots.
- Nearby precursor flag labels to quickly create an average or multi-spectrum for all dependent acquisitions.
- Customizable number formats to modify decimal and list separators to match different country localizations.
- Display of mass spectrometry trailer (MS Trending) information as a function of time.
- Multiple chromatograms can be overlaid for easier comparison.