The following table lists known defects in the software, which are categorized by software section. Each defect row includes a brief abstract about the defect and information related to each defect's severity and risk. The Item ID corresponds to the internal number assigned to each issue. Product management assesses the risk based on the criteria listed above, which can differ significantly from the reported severity.
Software section | Item ID | Abstract | Severity | Risk |
|---|---|---|---|---|
Data Viewer | 399523 | The Average a Spectrum operation is not removed from the Undo list after performing an Undo action (affects only the user interface; the data remains correct). This occurs when multiple spectra are added and removed, and then multiple undo actions are performed. This leads to the exclusion of averaging from the undo actions. Workaround: To remove the averaging action, either undo the actions step by step or select the spectra. | Medium | Low |
Data Viewer | 360190 | The pump pressure channel displayed under the Detector type and Detector channels are not in sequence (only for Chromeleon data). | Medium | No Risk |
Data Viewer | 751036 | The Undo action does not restore default spectrum size. | Medium | No Risk |
Data Viewer | 751016 | The Multi Spectrum trace is not updated after changing the Scan filter. | Medium | Low |
Data Viewer | 751049 | The Undo/Redo functionality is not consistent for precursor ion generation (by using the Precursor flag). | Medium | Low |
Data Viewer | 751050 | Averaging is performed in a different trace when an MS trending trace is present. | Medium | Medium |
Data Viewer | 751051 | The Plot properties display four decimal value in Mass Range field | Low | Low |
Data Viewer | 748016 | Spectrum scan data is not updated after performing an undo operation in a newly inserted spectrum. | Medium | Medium |
Data Viewer | 753976 | Users can open injections with an upload status of “Raw File Validation Error” directly from the Object Picker. However, opening files with an upload status of Failed or Waiting will trigger an error in the Data Viewer application. | Low | Low |