You can access the shortcut menu of a Spectrum view or a MultiSpectrum view by right-clicking the view.
Command | Description |
|---|---|
Insert Spectrum View | Adds a copy of the active spectrum. |
Display Options | Opens the Display Options dialog box for labeling the mass spectrum peaks. |
Export as Image | Copies an image of the Spectrum view to the clipboard or downloads the active Spectrum view as an SVG image. |
Reset Scaling | Resets the scaling of any plots in either a Spectrum view or a MultiSpectrum view. This command is only available when the spectrum is zoomed in. |