If you have multiple chromatogram traces open in a Chromatogram view, you can remove all but one trace. If only one trace is open, the Close icon
is unavailable.
NOTE
You can also remove a trace using the Ranges dialog box.
NOTE
You can also remove a trace using the Ranges dialog box.