You can access the Chromatogram view's shortcut menu by right-clicking the view.
Command | Description |
|---|---|
Average Spectrum | Create an average spectrum, which you do by dragging the cursor across the x-axis to choose an RT range. |
Export as Image | Copies an image of the Chromatogram view to the clipboard or downloads the active Chromatogram view as an SVG image. |
Ranges | Displays the Ranges tab that is linked to the active Chromatogram view. |
Reset Scaling | Resets the scaling of all the plots in the Chromatogram view. Enabled when chromatogram is zoomed in. |