The Import Mass List dialog box lets you import a CSV file and map the CSV header to the chromatogram ranges to create XIC traces in the active Chromatogram view. The application reads only files with a .csv file name extension. The following table describes the parameters in the Import Mass List dialog box.
Parameter | Description |
|---|---|
Header Row | Specifies the row number of the CSV file that contains CSV headers. Default: 1. |
File Name | Specifies the path and file name of the CSV file to be imported. |
Filter | Specifies a filter type of the XIC to apply to the file. |
Mass Tolerance | Specifies the mass tolerance for the m/z of the specified ion. Available only when you select Mass Range under Trace Type. Default mass tolerance for different mass analyzer types:
Range: 0.00−1,000 ppm or 0.00−10 amu; units: amu, mmu, or ppm. Default: 5 ppm. |
Units | Specifies the following units for the Mass Tolerance parameter:
Default: ppm. |
Column Separator | Specifies a character used to separate columns in a data file based on a CSV format, such as:
By default, the column separator is derived from the User Profile Settings. |
Replace Existing Table | Replaces the existing traces with new traces corresponding to the mapped CSV values. |
Append Existing Table | Adds the new traces corresponding to the mapped CSV values with the existing traces. |
CSV Header- Ranges | Enables the CSV header element to map to the corresponding Ranges parameter. |
CSV Header- Compound Name Label | Enables the CSV header element to map to the corresponding Compound Name parameter. |