The Chromatogram view displays the chromatogram trace for the raw data file. The x axis displays the time (minutes), and the y axis displays the relative abundance. Users can zoom in on, reset, and modify the Chromatogram view through the ellipsis menu .
TIP
You can resize the Chromatogram view by dragging the splitter bar that is positioned between the Chromatogram view and the Spectrum view; dragging the bar up decreases the Chromatogram view's width and dragging it down increases the view's width.