Users can zoom in on and navigate the Chromatogram view using:
  • The mouse pointer
  • The Zoom Range option in the Processing menu
The application updates the RT range in the header and the NL value in the title as users zoom in on different RT regions.