Allows users to export the Chromatogram view by downloading it as an SVG file or by copying the view to the clipboard and then pasting it into a document. The following table lists the parameters in the Export as Image dialog box.
Parameter | Subparameter | Description |
---|---|---|
Copy to Clipboard | N/A | Copies the Chromatogram view to the clipboard, which the user can then paste into a document. |
Download as SVG | N/A | Saves the image as an SVG file to the local folder; the user can then insert the file into a document. |
Dimensions | N/A | Specifies the width and height of the downloaded image. |
Width | Specifies the width of the image. Range: 2–50; units: user-specified. | |
Height | Specifies the height of the image. Range: 2–50; units: user-specified. | |
Units | Displays the following units of the dimension:
|