The Ranges page allows users to add, delete, import, and define the chromatogram traces displayed in the Chromatogram view.
Function | Description |
|---|---|
Add Trace | Adds a new row with the defined trace from the previously selected row. |
Import Mass List | Imports a CSV file and maps the CSV Header element to the Ranges element to create XIC traces in the active Chromatogram view. For more information, see Import a CSV file. |
Remove | Removes the selected trace from the list of chromatogram ranges. |
Reset | Restores the chromatogram ranges parameters. |
Column | Rearranges the columns that appear in the Ranges dialog box. |
Trace visibility | Displays ( |
Plot Properties and Processing | Define different parameters of traces. For more information, see Plot Properties and Processing. |