The following table defines the parameters used to calculate the S/N using the Pharmacopeia S/N method.
Parameter | Subparameter | Description |
---|---|---|
Pharmacopoeia S/N Calculation | Checkbox | Specifies a calculation that overwrites the S/N calculation from ICIS peak detection settings. |
Time Span Multiply Factor | N/A | Specifies the factor (n) for multiplying the PWHH to calculate the time interval. |