The application saves an active workspace after a user signs out or exits from the Ardia Platform or the Data Viewer application, but only if the user had performed a function (like modifying a Chromatogram view or Spectrum view) in the workspace before exiting or signing out.
The application restores a saved workspace when the user restarts or logs in to it again.