The following table lists the icons in the workspace that provide users with quick access to different functions.
Icons
Description
Undo icon
Cancels the previous 20 actions. The button is active only after user performing the following actions in the active pane:
  • Scan Selection in Trace: Navigates across the scans and change the RT in the Chromatogram view.
  • Zooming Chromatogram View: Zooms in on a Chromatogram view.
  • Reset Chromatogram View: Resets a zoomed Chromatogram view.
  • Zooming Spectrum View: Zooms in on a Spectrum view.
  • Reset Spectrum View: Resets a zoomed Spectrum view.
  • Chromatogram Ranges Change: Adds, deletes, or modifies chromatogram traces in the Chromatogram view from the Ranges dialog box.
  • Peak Detection: Identify and integrate the chromatogram peaks in an active chromatogram plot from the Peak Detection page.
  • Spectrum Display Options Change: Adds or removes labels that appear above the spectrum peaks.
  • Averaging: Adds Average Spectrum over an appropriate RT range or scan range.
Redo icon
Reverses the previous 20 actions. The button is active after performing an undo action.
Expand icon
Expands the menu command pane. This command is only available when the menu command pane is collapsed.
Collapse icon
Collapses the menu command pane. This command is only available when the menu command pane is expanded.
icon
Displays the Ardia repository to browse the data file or injections.
icon
Displays the Info Bar where users can set the Peak Detection parameter and the Ranges parameter. To hide the Info Bar, turn off the toggle icon. For more information, see Info Bar.