The following functions allow users to process chromatograms.
Function | Subfunction | Description |
|---|---|---|
Average Spectrum | N/A | Activates the Average Spectra mode to improve mass accuracy and reduce noise in a spectrum. Users can also access the Average spectrum from the ellipsis menu |
Scan Range | Averages mass spectra based on the scan number or the range of scan numbers that users enter. | |
RT Range | Averages mass spectra based on the retention time (RT) or the range of RTs that users enter. | |
Zoom Range | Radio button | Zooms in to a part of the mass spectrum (or mass spectra) based on the RT range entered. |
NOTE
The list and decimal separators are determined by either your browser's regional settings or your preferences in User Settings. Refresh the page to view the applied changes. On refresh, any unapplied or unsaved changes will be lost. For more information, refer Change the list and decimal separators in the Ardia Core Software User Guide.