The File menu contains the following commands.
Command | Description |
|---|---|
Open | Opens the Data Explorer window where users can browse for data files or injections in the Ardia repository. |
Help | Downloads the Ardia Data Viewer Application User Guide as a PDF file. |
About | Displays the version information, including the application name, build version, and build date. |