The Thermo Scientific™ Ardia™ Data Viewer application is a web-based application that lets users visualize chromatography and MS data. Users can access injections or raw data files from the Ardia repository, which are collected through the Xcalibur™ data system or the Chromeleon™ Chromatography Data System (CDS). The application allows users to display Chromatogram views and Spectrum views, navigate the scans, generate extracted ion chromatograms (XICs), and view chromatogram traces from multiple detectors and multiple raw data files.