The following table lists known defects in the software, which are categorized by software section. Each defect row includes a brief abstract about the defect and information related to each defect's severity and risk. The Item ID corresponds to the internal number assigned to each issue. Product management assesses the risk based on the criteria listed above, which can differ significantly from the reported severity.
Software section | Item ID | Abstract | Severity | Risk |
|---|---|---|---|---|
Data Viewer | 399523 | The Average a Spectrum operation is not removed from the Undo list after performing an Undo action (affects only the user interface; the data remains correct). This occurs when multiple spectra are added and removed, and then multiple undo actions are performed. This leads to the exclusion of averaging from the undo actions. Workaround: To remove the averaging action, either undo the actions step by step or select the spectra. | Medium | Low |
Data Viewer | 393343 | Diagonal zooming is not performed with the labeled view, and the zooming operation does not support the Undo operation. | Medium | Low |
Data Viewer | 360190 | The pump pressure channel displayed under the Detector type and Detector channels are not in sequence (only for Chromeleon data) | Medium | No Risk |