The Data Viewer application allows users to do the following:
- Access Chromeleon injections. This includes both 2D and MS-based injections. For details on synchronizing Chromeleon data vaults with the Ardia Platform, refer to the Chromeleon Help.
- Access MS data from the Ardia repository (including the new Thermo Scientific Orbitrap™ Astral™ Mass Spectrometer data) through the Data Viewer and Data Explorer. This applies to data uploaded manually or automatically via the Xcalibur software.
- Restore and reapply up to the previous 20 actions using the Undo and Redo buttons.
- Zoom in on the Chromatogram view and the Spectrum view.
- Restore any zoomed action using the Reset Zoom button to display the full Chromatogram view or Spectrum view.
- Save an active workspace by exiting or signing out of the Ardia Platform or the Data Viewer application.
- Restore the saved workspace by restarting or signing in to the application.
- Generate XICs by manually entering a value in the Ranges dialog box or double-clicking the m/z value in the Spectrum view.
- Calculate the m/z for a trace by entering a chemical formula or a peptide sequence (single-letter code).
- Import a CSV file with m/z value and compound name (optional) to append or replace the chromatogram ranges table and subsequently generate XICs (up to 50).
- Identify and integrate chromatogram peaks into a single trace or multiple traces using ICIS or Genesis peak detection algorithms.
- Apply the Pharmacopeia Signal-to-Noise (S/N) calculation to overwrite the S/N calculation from the ICIS peak detection using the Pharmacopeia formula.
- Export a peak list as a CSV file and save it locally. The peak list is displayed as a table of peaks in the integrated chromatogram traces.
- Apply custom labels to the spectrum and chromatogram peaks to display them on the respective views.
- Open data-dependent scans in another Spectrum view using the Precursor Flag marker and the Nearby Precursors marker.
- Add up to ten multiple-spectrum views to the workspace and then link to a chromatogram trace.
- Exports a Spectrum view to other applications, such as Thermo Scientific Mass Frontier™, Thermo Scientific myLibrary™ Enterprise, or Thermo Scientific mzCloud™.
- Export a Chromatogram view and a Spectrum view as images or SVG files and copy them to the clipboard. Users can paste the images into an appropriate document.
- Average a spectrum over an appropriate RT range, improving mass accuracy and reducing noise in the spectrum.