In the Audit Trail Viewer application, you can sort the contents of the following columns:
Procedure
- Open the Audit Trail Viewer.
- Select the header of the column that you want to sort.
- The application sorts the list in ascending or chronological order based on the column's contents, as shown in the following table.
Column | Sort type |
---|---|
Timestamp | Chronological |
Object Type | Alphabetical |
Object Name | Alphanumerical |
Application | Alphabetical |
Action | Alphabetical |
Username | Alphabetical |
Role | Alphabetical |
Comment | Alphanumerical |
Path | Alphanumerical |
Additional Info | Alphabetical |
- (Optional) Select the header again to reverse the sort order.