You can adjust the column width in the Audit Trail Viewer application to fit your requirements.
Procedure
  1. Point to the right side of a column boundary until the resize cursor appears.
  2. Drag the column boundary to adjust the width to your preference.