In the Data Explorer application, you can pin your frequently used folders and sequences to the Quick Access area.
Prerequisites
- Your role must have the required Ardia Core permission to open the Data Explorer application. For more information, see Edit Ardia Core privileges for a role.
Procedure
- Open the Data Explorer application.
- In the navigation pane, navigate to the folder or the sequence you want to pin to the Quick Access area.
- Right-click the folder and then select Pin to Quick Access.
- The folder is listed in the Quick Access area in the navigation pane, with a pin icon next to the folder name.
- (Optional) To remove a folder from the Quick Access, right-click the folder and then select Remove from Quick Access.
NOTE
The recently created folders are also listed in the Quick Access area.