From the Data Explorer application, you can open injections and raw data files directly in the Ardia Data Viewer.
Prerequisites
- Your role must have the required Ardia Core permission to open the Data Explorer application. For more information, see Edit Ardia Core privileges for a role.
- You must have the Can View permission (or a higher permission) for the folder containing the injections or the raw data files you want to open. For more information, see Manage folder access.
Procedure
- Open the Data Explorer application.
- In the content area, do one of the following:
- Select one or more injections or raw data files and then select Open on the command bar.
NOTE
You can open a maximum of 15 injections in the Data Viewer. If you select more that 15 injections, the Open command is unavailable.
- Select on an injection or a raw data file row and then select Open.
- Right-click an injection or a raw data file row and then select Open.
- The Data Viewer application opens in a new tab with each selected file open in a separate workspace.