Connected applications, such as BioPharma Finder or Proteome Discoverer, require one or more raw data files as the input for every experiment, no matter the workflow. Such raw data files contain your experimental data and are processed by the application according to your selected processing method.
Using the Data Explorer application, you can upload raw data files to the Ardia Repository. The Ardia Repository stores the uploaded raw data files within the sequence created by the application. A sequence can contain multiple raw data files. You can upload a raw data file to a new sequence or an existing sequence.
With the appropriate permissions, you can: